Learn More
Ansell™ RINGERS™ R668
Catalog No. 17720637
Click to view available options
Size:
8
9
10
11
12
13
Premium leather with 360° cut, level 2 impact and G-100 Thinsulate™ for cold weather.
  • Thermal goatskin leather: G-100 Thinsulate lined for warmth and durability
  • Superior cut-resistant lining: For impact gloves with 360° cut resistance
  • Patented TPR design: Knuckles, thumb and finger-length impact protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Cut A6, ANSI ABR 4, ANSI/ISEA 105-2006 PUN 3
Coated Area Uncoated
Construction Cut and Sewn
Description Medium-duty impact gloves are fully lined with high-quality goatskin leather.
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material HPPE
Product Type Mechanical Protection Gloves
Size 10
Gender Unisex
Glove Cuff Color White
Glove Cuff Style Open
Grip Design Goatskin leather
Liner Material HPPE, Thinsulate, Polyester, Goatskin leather
Lining Lined
Model R668
Puncture Rating ANSI/ISEA 105-16 Level 3
Thumb Style Wing
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.