Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RINL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545964
Description
Peptide sequence: ETHRGWGREQ TPQETEPEAA QRHDPAPRNP APHGVSWVKG PLSPEVDHPG Sequence homology: Human: 100%.
Guanine nucleotide exchange factor (GEF) for RAB5A and RAB22A that activates RAB5A and RAB22A by exchanging bound GDP for free GTP. Plays a role in endocytosis via its role in activating Rab family members.
Specifications
RINL | |
Polyclonal | |
Unconjugated | |
RINL | |
Ras and Rab interactor like; Ras and Rab interactor-like; ras and Rab interactor-like protein; RINL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
126432 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZS11 | |
RINL | |
Synthetic peptide directed towards the N-terminal of human RINL (aa 176-225). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction