Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RINL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559860 Supplier Invitrogen™ Supplier No. PA559860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDVLPRTLLL PPPTLGPRDE HTDPVQIGRV QQDTPGKVLS IVNQLYLETH RGWGREQTPQ ETEPEAAQRH DPAPRNPAPH GVSWVKGPLS Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 63%.

Guanine nucleotide exchange factor (GEF) for RAB5A and RAB22A that activates RAB5A and RAB22A by exchanging bound GDP for free GTP. Plays a role in endocytosis via its role in activating Rab family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RINL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RINL
Gene Accession No. Q6ZS11
Gene Alias Ras and Rab interactor like; Ras and Rab interactor-like; ras and Rab interactor-like protein; RINL
Gene Symbols RINL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RINL. Recombinant protein control fragment (Product #RP-99190).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126432
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.