Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RINT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554104 Supplier Invitrogen™ Supplier No. PA554104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PCCSESGDER KNLEEKSDIN VTVLIGSKQV SEGTDNGDLP SYVSAFIEKE VGNDLKSLKK LDKLIEQRTV SKMQLEEQVL TISSEIPKRI RSALKNAEES KQFLNQFLEQ ETHLFS Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 68%.

Involved in regulation of membrane traffic between the Golgi and the endoplasmic reticulum (ER); the function is proposed to depend on its association in the NRZ complex which is believed to play a role in SNARE assembly at the ER. May play a role in cell cycle checkpoint control. Essential for telomere length control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RINT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RINT1
Gene Accession No. Q6NUQ1
Gene Alias 1500019C06Rik; 2810450M21Rik; hsRINT-1; RAD50 interactor 1; RAD50-interacting protein 1; RGD1560433; RINT1; RINT-1
Gene Symbols RINT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RINT1. Recombinant protein control fragment (Product #RP-92725).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60561
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.