Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIOK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562409 Supplier Invitrogen™ Supplier No. PA562409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DWDWDEGVGK LAKGYVWNGG SNPQANRQTS DSSSAKMSTP ADKVLRKFEN KINLDKLNVT DSVINKVTEK SRQKEADMYR IKD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.

FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predominantly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIOK1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Riok1
Gene Accession No. Q9BRS2
Gene Alias 3110046C13Rik; 5430416A05Rik; AD034; bA288G3.1; RIO kinase 1; RIO kinase 1 (yeast); RIO1; Riok1; RRP10; Serine/threonine-protein kinase RIO1
Gene Symbols Riok1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RIOK1. Recombinant protein control fragment (Product #RP-99573).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83732
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.