Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIOK2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130098JF549
Description
RIOK2 Polyclonal antibody specifically detects RIOK2 in Human samples. It is validated for Western BlotSpecifications
RIOK2 | |
Polyclonal | |
Western Blot | |
EC 2.7.11.1, FLJ11159, RIO kinase 2, RIO kinase 2 (yeast), RIO2, serine/threonine-protein kinase RIO2 | |
Synthetic peptide mapping to a region between residue 275 and 325 of human RIO kinase 2 using the numbering given in entry NP_060813.1 (GeneID 55781). | |
0.1 mL | |
Protein Kinase | |
55781 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction