Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIOK2 Antibody (OTI3E11), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273910AF647
Description
RIOK2 Monoclonal specifically detects RIOK2 in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
RIOK2 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RIOK2 | |
Recombinant protein fragment corresponding to amino acids 277-552 of human RIOK2 (NP_060813) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3E11 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11.1, FLJ11159, RIO kinase 2, RIO kinase 2 (yeast), RIO2, serine/threonine-protein kinase RIO2 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
55781.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction