Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIOK2 Monoclonal Antibody (OTI3E11)
GREENER_CHOICE

Catalog No. PIMA526060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526060 Supplier Invitrogen™ Supplier No. MA526060
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RIOK2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

RIOK2 belongs to the protein kinase superfamily; RIO-type Ser/Thr kinase family. Serine/threonine protein kinases, such as RIOK2, phosphorylate the OH group of serine or threonine (which have similar sidechains). Activity of these protein kinases can be regulated by specific events (e.g. DNA damage), as well as numerous chemical signals, including cAMP/cGMP, Diacylglycerol, and Ca2+/calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIOK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RIOK2
Gene Accession No. Q9BVS4, Q9CQS5
Gene Alias 2010110K24Rik; 2410085M17Rik; RIO kinase 2; RIO kinase 2 (yeast); RIO2; Riok2; Serine/threonine-protein kinase RIO2
Gene Symbols RIOK2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 277-552 of RIOK2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55781, 67045
Target Species Human, Mouse, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.