Learn More
RIOK2 Mouse anti-Human, Mouse, Clone: OTI3E11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505140

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RIOK2 belongs to the protein kinase superfamily; RIO-type Ser/Thr kinase family. Serine/threonine protein kinases, such as RIOK2, phosphorylate the OH group of serine or threonine (which have similar sidechains). Activity of these protein kinases can be regulated by specific events (e.g. DNA damage), as well as numerous chemical signals, including cAMP/cGMP, Diacylglycerol, and Ca2+/calmodulin.Specifications
RIOK2 | |
Monoclonal | |
Unconjugated | |
RIOK2 | |
RIO2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55781, 67045 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3E11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BVS4, Q9CQS5 | |
RIOK2 | |
Human recombit protein fragment corresponding to amino acids 277-552 of human RIOK2 produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.