Learn More
Invitrogen™ RIOK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114100
Description
Immunogen sequence: IDYNRHAVVM ELINGYPLCQ IHHVEDPASV YDEAMELIVK LANHGLIHGD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
RIOK2 belongs to the protein kinase superfamily; RIO-type Ser/Thr kinase family. Serine/threonine protein kinases, such as RIOK2, phosphorylate the OH group of serine or threonine (which have similar sidechains). Activity of these protein kinases can be regulated by specific events (e.g. DNA damage), as well as numerous chemical signals, including cAMP/cGMP, Diacylglycerol, and Ca2+/calmodulin.
Specifications
| RIOK2 | |
| Polyclonal | |
| Unconjugated | |
| RIOK2 | |
| 2010110K24Rik; 2410085M17Rik; RIO kinase 2; RIO kinase 2 (yeast); RIO2; Riok2; Serine/threonine-protein kinase RIO2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55781 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BVS4 | |
| RIOK2 | |
| Synthetic peptide directed towards the middle region of human RIOK2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.