Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIOK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551615 Supplier Invitrogen™ Supplier No. PA551615
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IGKGKDITTK HDEVVCGRKN TARMENFAPE FQVGDGIGMD LKLSNHVFNA LKQHAYSEER RSARLHEKKE HSTAEKAVDP KTRSLMYKMV NSGMLETITG CISTGKESVV FHAYGGSMED EKEDSKVIPT ECA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. SUDD, also known as RIOK3 (RIO kinase 3), is a 519 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Expressed in a variety of tissues, SUDD catayzes the ATP-dependent phosphorylation of target proteins, thereby influencing signaling events throughout the cell. SUDD is expressed as two isoforms due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIOK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RIOK3
Gene Accession No. O14730
Gene Alias 1200013N13Rik; D18Ertd331e; E130306C24Rik; homolog of the Aspergillus nidulans sudD gene product; RIO kinase 3; RIOK3; Serine/threonine-protein kinase RIO3; SUDD; sudD (suppressor of bimD6, Aspergillus nidulans) homolog; sudD homolog; sudD suppressor of Aspergillus nidulans bimD6 homolog; sudD suppressor of bimD6 homolog; sudD, suppressor of bimD6 homolog; testicular secretory protein Li 47
Gene Symbols RIOK3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RIOK3. Recombinant protein control fragment (Product #RP-101005).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8780
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.