Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIOK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528585 Supplier Invitrogen™ Supplier No. PA528585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HepG2, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (94%), Rat (94%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. SUDD, also known as RIOK3 (RIO kinase 3), is a 519 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Expressed in a variety of tissues, SUDD catayzes the ATP-dependent phosphorylation of target proteins, thereby influencing signaling events throughout the cell. SUDD is expressed as two isoforms due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIOK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RIOK3
Gene Accession No. O14730, Q9DBU3
Gene Alias 1200013N13Rik; D18Ertd331e; E130306C24Rik; homolog of the Aspergillus nidulans sudD gene product; RIO kinase 3; RIOK3; Serine/threonine-protein kinase RIO3; SUDD; sudD (suppressor of bimD6, Aspergillus nidulans) homolog; sudD homolog; sudD suppressor of Aspergillus nidulans bimD6 homolog; sudD suppressor of bimD6 homolog; sudD, suppressor of bimD6 homolog; testicular secretory protein Li 47
Gene Symbols RIOK3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RIOK3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361293, 66878, 8780
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.