Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIOK3 Polyclonal Antibody
RIOK3 Polyclonal Antibody
Supplier: Thermo Scientific PA537066
Description
RIOK3 Polyclonal Antibody for Western Blot

Specifications
RIOK3 | |
Polyclonal | |
Unconjugated | |
RIOK3 | |
1200013N13Rik; D18Ertd331e; E130306C24Rik; homolog of the Aspergillus nidulans sudD gene product; RIO kinase 3; RIOK3; Serine/threonine-protein kinase RIO3; SUDD; sudD (suppressor of bimD6, Aspergillus nidulans) homolog; sudD homolog; sudD suppressor of Aspergillus nidulans bimD6 homolog; sudD suppressor of bimD6 homolog; sudD, suppressor of bimD6 homolog; testicular secretory protein Li 47 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361293, 66878, 8780 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide | |
O14730, Q9DBU3 | |
RIOK3 | |
Synthetic peptide corresponding to amino acids 322-367 of human RIOK3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction