Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIP1 Monoclonal Antibody (C4), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA550961
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550961 200 μL
FEMA550962 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550961 Supplier Invitrogen™ Supplier No. MA550961
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serine-threonine kinase which transduces inflammatory and cell-death signals (programmed necrosis) following death receptors ligation, activation of pathogen recognition receptors (PRRs), and DNA damage. Upon activation of TNFR1 by the TNF-alpha family cytokines, TRADD and TRAF2 are recruited to the receptor. Phosphorylates DAB2IP at 'Ser-728' in a TNF-alpha-dependent manner, and thereby activates the MAP3K5-JNK apoptotic cascade. Ubiquitination by TRAF2 via 'Lys-63'-link chains acts as a critical enhancer of communication with downstream signal transducers in the mitogen-activated protein kinase pathway and the NF-kappa-B pathway, which in turn mediate downstream events including the activation of genes encoding inflammatory molecules. Polyubiquitinated protein binds to IKBKG/NEMO, the regulatory subunit of the IKK complex, a critical event for NF-kappa-B activation. Interaction with other cellular RHIM-containing adapters initiates gene activation and cell death. RIPK1 and RIPK3 association, in particular, forms a necrosis-inducing complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIP1
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene RIPK1
Gene Accession No. Q13546
Gene Alias cell death protein RIP; D330015H01Rik; FLJ39204; OTTHUMP00000015955; receptor (TNFRSF)-interacting serine-threonine kinase 1; receptor interacting serine/threonine kinase 1; receptor-interacting protein 1; receptor-interacting protein kinase 1; receptor-interacting serine/threonine-protein kinase 1; Rinp; RIP; RIP1; RIP-1; RIP1 (CT); RIPK1; serine/threonine-protein kinase RIP
Gene Symbols RIPK1
Host Species Mouse
Immunogen Recombinant Human RIPK1 protein, Phe17-Tyr289 (Accession #Q13546).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100524751, 8737
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.