Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIP140 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1105122
Description
RIP140 Polyclonal specifically detects RIP140 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
RIP140 | |
Polyclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
NRIP1 | |
Synthetic peptide corresponding to residues G(718) N P T K G R V K K K E K T P L R D(735) of human RIP140. | |
Affinity Purified | |
RUO | |
Primary | |
Detects a band at approximately 140 kDa. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1 mg/mL | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
10RIP140Receptor-interacting protein 140, FLJ77253, nuclear receptor interacting protein 1, nuclear receptor-interacting protein 1, receptor interacting protein 140 | |
Rabbit | |
140 kDa | |
100 μg | |
Transcription Factors and Regulators | |
8204 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction