Learn More
Invitrogen™ RIP3 Monoclonal Antibody (Rippy-3)

Description
Predicted molecular weight of MA1-25841 is 62 kDa.
Specifications
Specifications
| Antigen | RIP3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | Rippy-3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RIPK3 |
| Gene Accession No. | Q9Y572 |
| Gene Alias | 2610528K09Rik; AW107945; Hcyp2; Homocysteine respondent protein HCYP2; mRIP3; receptor interacting protein 3; receptor interacting serine/threonine kinase 3; receptor-interacting protein 3; receptor-interacting serine/threonine-protein kinase 3; receptor-interacting serine-threonine kinase 3; Rip3; RIP-3; Ripk3; RIP-like protein kinase 3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.