Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RIPA 56
GREENER_CHOICESDP

Catalog No. 733950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-395-0 50 mg
73-391-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 73-395-0 Supplier Tocris Bioscience™ Supplier No. 7339/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and metabolically stable RIP1 kinase inhibitor

RIPA 56 is a potent and metabolically stable receptor-interacting protein kinase 1 (RIPK1) inhibitor (EC50 = 13 nM). RIPA-56 shows no inhibition of RIP3 kinase at 10 μM . RIPA 56 blocks necroptosis and aging of the mouse male reproductive system. RIPA 56 reduces tumor necrosis factor α (TNFα)-induced mortality and multi-organ damage in a mouse model of systemic inflammatory response syndrome (SIRS). RIPA 56 is orally bioavailable.

Specifications

Chemical Name or Material N-Hydroxy-2,2-dimethyl-N-(phenylmethyl)butanamide
CAS 1956370-21-0
Quantity 50 mg
Target RIP Kinase Inhibitors
Molecular Formula C13H19NO2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.