Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPA Buffer, 5X, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Description
General Description
- RIPA can be used for isolation of cytoplasmic, membrane, nuclear, and mitochondrial proteins
- Protease inhibitors can be added to RIPA buffer to assist in protein isolation
Application
- RIPA buffer is suitable for use in protein isolation from culture cells or homogenized tissues
- Downstream applications of RIPA buffer include ELISA, western blot, immunoprecipitation, and SDS-PAGE

Specifications
Specifications
Quantity | 100 mL |
Format | Liquid |
Product Type | Cell Lysis Reagent |
Concentration | 5X |
Content And Storage | 2° to 8°C |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction