Learn More
Thermo Scientific™ RIPA Lysis and Extraction Buffer, 100 mL PROMO
Description
This RIPA buffer effectively lyses and extracts protein from cultured mammalian cells, including plated cells and pelleted suspension cells. The popular reagent enables the extraction of membrane, nuclear and cytoplasmic proteins and is compatible with many applications, including reporter assays, the Thermo Scientific BCA Protein Assay, immunoassays and protein purification. Inhibitors such as Thermo Scientific Halt Protease Inhibitor Cocktail (Part No. 78430) and Halt Phosphatase Inhibitor Cocktail (Part No. 78420) are also compatible with this RIPA buffer formulation and can be added before use to prevent proteolysis and maintain protein phosphorylation.
Highlights:
Convenient – ready-to-use solution
Flexible – compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification
Versatile – enables extraction of cytoplasmic, membrane and nuclear proteins
Disclosed formulation – contains no proprietary components, providing users with complete control and knowledge of possible compatibility issues
Specifications
Specifications
| Quantity | 100 mL |
| Format | Liquid |
| Product Type | Extraction Buffer |
| Volume (Metric) | 100 mL |
| Content And Storage | Upon receipt store at 4°C. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.