Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RIPA Lysis and Extraction Buffer PROMO

Catalog No. PI89901PM
Click to view available options
:
Thermo Scientific™ RIPA Lysis and Extraction Buffer

Catalog No. PI89901PM

Supplier: Thermo Scientific™ 89901

Lyse cultured mammalian cells with this high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent.

This RIPA buffer effectively lyses and extracts protein from cultured mammalian cells, including plated cells and pelleted suspension cells. The popular reagent enables the extraction of membrane, nuclear and cytoplasmic proteins and is compatible with many applications, including reporter assays, the Thermo Scientific BCA Protein Assay, immunoassays and protein purification. Inhibitors such as Thermo Scientific Halt Protease Inhibitor Cocktail (Part No. 78430) and Halt Phosphatase Inhibitor Cocktail (Part No. 78420) are also compatible with this RIPA buffer formulation and can be added before use to prevent proteolysis and maintain protein phosphorylation.

Highlights:

  • Convenient – ready-to-use solution

  • Flexible – compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification

  • Versatile – enables extraction of cytoplasmic, membrane and nuclear proteins

  • Disclosed formulation – contains no proprietary components, providing users with complete control and knowledge of possible compatibility issues

Specifications

Quantity 250 mL
Format Liquid
Product Type Extraction Buffer
Volume (Metric) 250 mL
Content And Storage Upon receipt store at 4°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.