Learn More
RIPK1 Mouse anti-Canine, Human, Clone: OTI1A7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800328

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Serine-threonine kinase which transduces inflammatory and cell-death signals (programmed necrosis) following death receptors ligation, activation of pathogen recognition receptors (PRRs), and DNA damage. Upon activation of TNFR1 by the TNF-alpha family cytokines, TRADD and TRAF2 are recruited to the receptor. Phosphorylates DAB2IP at ′Ser-728′ in a TNF-alpha-dependent manner, and thereby activates the MAP3K5-JNK apoptotic cascade. Ubiquitination by TRAF2 via ′Lys-63′-link chains acts as a critical enhancer of communication with downstream signal transducers in the mitogen-activated protein kinase pathway and the NF-kappa-B pathway, which in turn mediate downstream events including the activation of genes encoding inflammatory molecules. Polyubiquitinated protein binds to IKBKG/NEMO, the regulatory subunit of the IKK complex, a critical event for NF-kappa-B activation. Interaction with other cellular RHIM-containing adapters initiates gene activation and cell death. RIPK1 and RIPK3 association, in particular, forms a necrosis-inducing complex.Specifications
RIPK1 | |
Monoclonal | |
Unconjugated | |
RIPK1 | |
RIP, RIP-1, RIP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
102152063, 8737 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1A7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13546 | |
RIPK1 | |
Human recombit protein fragment corresponding to amino acids 133-422 of human RIPK1 produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.