Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPK1/RIP1 Antibody (CL11357), Novus Biologicals™

Mouse Monoclonal Antibody
$330.00 - $547.00
Specifications
Antigen | RIPK1/RIP1 |
---|---|
Clone | CL11357 |
Dilution | Western Blot 1 ug/ml |
Applications | Western Blot |
Classification | Monoclonal |
Description
RIPK1/RIP1 Monoclonal antibody specifically detects RIPK1/RIP1 in Human samples. It is validated for Western BlotSpecifications
RIPK1/RIP1 | |
Western Blot 1 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of Q13546. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL11357 | |
Western Blot | |
Unconjugated | |
Mouse | |
Apoptosis, Necroptosis | |
PBS, pH 7.2, 40% glycerol | |
8737 | |
IgG2a | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title