Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK1/RIP1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Specifications

Antigen RIPK1/RIP1
Concentration 0.5 mg/mL
Dilution Western Blot 1 - 3 ug/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224739SS
SDP
View Documents
Novus Biologicals
NBP224739SS
0.025 mg N/A N/A N/A
NBP224739
SDP
View Documents
Novus Biologicals
NBP224739
0.1 mg N/A N/A N/A
Description

Description

RIPK1/RIP1 Polyclonal specifically detects RIPK1/RIP1 in Human, Primate, Rhesus Macaque samples. It is validated for Western Blot.
Specifications

Specifications

RIPK1/RIP1
Western Blot 1 - 3 ug/mL
Polyclonal
Purified
RUO
PBS, 0.2% Gelatin with 0.05% Sodium Azide
Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP
RIPK1
IgG
Protein G purified
The amino acid sequence used as immunogen is 100% homologous in human (multiple isoforms), rhesus monkey (multiple isoforms), and orangutan, 94% homologous in bovine, dog and rat, and 88% homologous in mouse.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Human
Q13546
8737
A portion of amino acids 350-400 of human RIP1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.