Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPK2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132424F
Description
RIPK2 Polyclonal specifically detects RIPK2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RIPK2 | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
FITC | |
CARD3, CARD-containing IL-1 beta ICE-kinase, CARD-containing interleukin-1 beta-converting enzyme (ICE)-associated kinase, CARD-containing interleukin-1 beta-converting enzyme-associated kinase, CARDIAKCCK, EC 2.7.11, EC 2.7.11.1, growth-inhibiting gene 30, receptor interacting protein 2, receptor-interacting protein (RIP)-like interacting caspase-like apoptosisregulatory protein (CLARP) kinase, Receptor-interacting protein 2, receptor-interacting serine/threonine-protein kinase 2, receptor-interacting serine-threonine kinase 2, RICKGIG30, RIP-2, RIP2CARD-carrying kinase, RIP-like-interacting CLARP kinase | |
Recombinant protein encompassing a sequence within the center region of human RIP2. The exact sequence is proprietary. | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
8767 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction