Learn More
Promega RIPK2 Kinase Enzyme System, 1mg
Catalog No. PRV6478 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6478 1 mg
1 options
Catalog No. PRV6478 Supplier Promega Supplier No. V6478
Add to Cart
Add to Cart
Request a Quote
RIPK2 (RIP2; RICK) is a death domain-containing protein kinase encoding a predicted 540-amino acid protein that contains an N-terminal serine/threonine kinase catalytic domain and a C-terminal caspase activation and recruitment domain.
  • Easily Screen and Profile RIPK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C