Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK3 Mouse anti-Human, Clone: OTI2F12, lyophilized, TrueMAB™
SDP

Catalog No. 501666358
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-6358 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-6358 Supplier OriGene Supplier No. CF803171
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

RIP3 is a novel member of RIP Ser/Thr kinase family. It is a potent inducer of apoptosis and is an important component of TNFR1 signaling complex. RIP3 consists of an N-terminal RIP-like-Kinase domain and a unique, non-homologous C-terminus responsible for caspase activation and apoptosis induction. RIP3 is expressed in multiple tissues including hematopoietic cells and plays a functional role in the regulation of apoptosis and NF-kB signaling. It is recruited to the TNFR1 signaling complex in a RIP-dependant manner where it induces apoptosis by activating caspases and/or inhibiting TNFR1-induced NF-kB activation. RIP3 negatively regulates the TLR3-Trif-mediated NF-kB signaling pathway by competing with the binding of Trif to RIP1. The human RIP3 gene is localized in the chromosomal region 14q11.2.

Specifications

Antigen RIPK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F12
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene RIPK3
Gene Accession No. Q9Y572
Gene Alias RIP3
Gene Symbols RIPK3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 233-518 of human RIPK3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11035
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.