Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPK3 Rabbit anti-Bovine, Human, Non-human Primate, Porcine, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc RIPK3301AP

Description
RIPK3 Polyclonal antibody specifically detects RIPK3 in Bovine, Human, Non-human Primate, Porcine samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
RIPK3 | |
Polyclonal | |
Unconjugated | |
RIPK3 | |
2610528K09Rik; AW107945; Hcyp2; Homocysteine respondent protein HCYP2; mRIP3; receptor interacting protein 3; receptor interacting serine/threonine kinase 3; receptor-interacting protein 3; receptor-interacting serine/threonine-protein kinase 3; receptor-interacting serine-threonine kinase 3; Rip3; RIP-3; Ripk3; RIP-like protein kinase 3 | |
Synthetic peptide taken within amino acid region 461-511 on human Receptor-interacting Serine/Threonine-protein kinase 3 protein. | |
200 μL | |
Primary | |
Bovine, Human, Non-human Primate, Pig | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q9Y572 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100153263, 11035, 452820, 507427 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction