Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK3/RIP3 Antibody (1312A) - BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$243.50 - $499.50

Specifications

Antigen RIPK3/RIP3
Clone 1312A
Concentration 1.0 mg/ml
Dilution Western Blot 2 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP276800SS
SDP
View Documents
Novus Biologicals
NBP276800SS
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP276800
SDP
View Documents
Novus Biologicals
NBP276800
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

RIPK3/RIP3 Monoclonal specifically detects RIPK3/RIP3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RIPK3/RIP3
1.0 mg/ml
Western Blot
Unconjugated
Rabbit
Human
EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
RIPK3
IgG
Protein A or G purified
56.7 kDa
1312A
Western Blot 2 ug/ml
Monoclonal
Purified
Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
PBS with 0.02% sodium azide
11035.0
RIPK3/RIP3 Antibody (1312A) was made to a partial synthetic peptide made to an internal portion of the human RIPK3/RIP3 protein (between amino acids 470-540). This seqeunce has 100% homology to isoform CRA_a and isoform CRA_b.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.