Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK3/RIP3 Antibody (1312D) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7161277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276799SS 0.025 mg
NBP276799 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP276799SS Supplier Novus Biologicals Supplier No. NBP276799SS
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RIPK3/RIP3 Monoclonal specifically detects RIPK3/RIP3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RIPK3/RIP3
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1312D
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 μ/mL, Immunocytochemistry/Immunofluorescence 5 - 15 μ/mL
Formulation PBS with 0.02% sodium azide
Gene Alias EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
Gene Symbols RIPK3
Host Species Rabbit
Immunogen RIPK3/RIP3 Antibody (1312D) was made to a partial synthetic peptide made to an internal portion of the human RIPK3/RIP3 protein (between amino acids 470-540). This seqeunce has 100% homology to isoform CRA_a and isoform CRA_b.
Molecular Weight of Antigen 56.7 kDa
Quantity 0.025 mg
Research Discipline Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11035
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.