Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ RIPK3/RIP3 Antibody Blocking Peptide

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 11035 |
Concentration | 1 mg/mL |
Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-24588. For further blocking peptide related protocol, click here . |
Gene Alias | EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 |
Gene Symbol | RIPK3 |
Quantity | 0.1 mg |
Research Category | Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction