Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPK3/RIP3 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232257UV
Description
RIPK3/RIP3 Polyclonal antibody specifically detects RIPK3/RIP3 in Human samples. It is validated for Western BlotSpecifications
RIPK3/RIP3 | |
Polyclonal | |
Western Blot | |
EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
RIPK3/RIP3 Antibody was made to a region between residue 400 to 450 of human Receptor Interacting Protein 3 using the numbering given in entry NP_006862.2 (GeneID 11035). | |
0.1 mL | |
Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction | |
11035 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction