Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIPK3/RIP3 Antibody (OTI1B3), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273912IR
Description
RIPK3/RIP3 Monoclonal specifically detects RIPK3/RIP3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| RIPK3/RIP3 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RIPK3 | |
| RIPK3/RIP3 Antibody (OTI1B3) [DyLight 755] was made to a recombinant protein fragment corresponding to amino acids 233-518 of human RIPK3/RIP3 (NP_006862) produced in E.coli. | |
| Affinity Purified | |
| Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction | |
| 11035.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1B3 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
| Mouse | |
| 56.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction