Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIPK4 Monoclonal Antibody (1F7D9)
GREENER_CHOICE

Catalog No. PIMA531895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531895 Supplier Invitrogen™ Supplier No. MA531895
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31895 has been tested in indirect ELISA.

The protein encoded by this gene is a serine/threonine protein kinase that interacts with protein kinase C-delta. The encoded protein can also activate NFkappaB and is required for keratinocyte differentiation. This kinase undergoes autophosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPK4
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1F7D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene RIPK4
Gene Accession No. P57078
Gene Alias 2310069J12Rik; AI552420; ANKK2; ANKRD3; ankyrin repeat domain 3; ankyrin repeat domain-containing protein 3; DIK; NKRD3; PKC-associated protein kinase; PKC-delta-interacting protein kinase; PKC-regulated protein kinase; pkk; PPS2; protein kinase C-associated kinase; protein kinase PKK; receptor interacting serine/threonine kinase 4; receptor-interacting serine/threonine-protein kinase 4; receptor-interacting serine-threonine kinase 4; RIP4; Ripk4; serine/threonine-protein kinase ANKRD3; wu:fj80b10; zgc:55705
Gene Symbols RIPK4
Host Species Mouse
Immunogen Purified recombinant fragment of human RIPK4 (AA: 675-784) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.