Learn More
Invitrogen™ RIPK4 Polyclonal Antibody

Description
RIPK4 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RIPK4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RIPK4 |
| Gene Accession No. | P57078, Q9ERK0 |
| Gene Alias | 2310069J12Rik; AI552420; ANKK2; ANKRD3; ankyrin repeat domain 3; ankyrin repeat domain-containing protein 3; DIK; NKRD3; PKC-associated protein kinase; PKC-delta-interacting protein kinase; PKC-regulated protein kinase; pkk; PPS2; protein kinase C-associated kinase; protein kinase PKK; receptor interacting serine/threonine kinase 4; receptor-interacting serine/threonine-protein kinase 4; receptor-interacting serine-threonine kinase 4; RIP4; Ripk4; serine/threonine-protein kinase ANKRD3; wu:fj80b10; zgc:55705 |
| Gene Symbols | RIPK4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.