Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502208882
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8882 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8882 Supplier Fabgennix Inc Supplier No. RIPK4401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RIPK4 Polyclonal antibody specifically detects RIPK4 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen RIPK4
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene RIPK4
Gene Accession No. P57078, Q9ERK0
Gene Alias 2310069J12Rik; AI552420; ANKK2; ANKRD3; ankyrin repeat domain 3; ankyrin repeat domain-containing protein 3; DIK; NKRD3; PKC-associated protein kinase; PKC-delta-interacting protein kinase; PKC-regulated protein kinase; pkk; PPS2; protein kinase C-associated kinase; protein kinase PKK; receptor interacting serine/threonine kinase 4; receptor-interacting serine/threonine-protein kinase 4; receptor-interacting serine-threonine kinase 4; RIP4; Ripk4; serine/threonine-protein kinase ANKRD3; wu:fj80b10; zgc:55705
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 250-300 on human RIPK4 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304053, 54101, 72388
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.