Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIPK5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550871

Catalog No. PA550871


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total RIPK5 / DSTYK protein.

RIPK5 is a serine/threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis. This gene encodes a dual serine/threonine and tyrosine protein kinase which is expressed in multiple tissues. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RIPK5
Polyclonal
Unconjugated
Dstyk
A930019K20Rik; C430014H23Rik; C820013G01; CAKUT1; Dstyk; Dual serine/threonine and tyrosine protein kinase; dusty PK; Dusty protein kinase; DustyPK; HDCMD38P; KIAA0472; receptor interacting protein kinase 5; receptor-interacting serine/threonine-protein kinase 5; RIP5; RIP-homologous kinase; Ripk5; sgK496; Sugen kinase 496
Rabbit
Antigen affinity chromatography
RUO
25778
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.3 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q6XUX3
Dstyk
Fusion protein corresponding to residues near the C terminal of human dual serine/threonine and tyrosine protein kinase.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.