Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIPK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551302 Supplier Invitrogen™ Supplier No. PA551302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total RIPK5 / DSTYK protein.

RIPK5 is a serine/threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis. This gene encodes a dual serine/threonine and tyrosine protein kinase which is expressed in multiple tissues. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPK5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Dstyk
Gene Accession No. Q6XUX3
Gene Alias A930019K20Rik; C430014H23Rik; C820013G01; CAKUT1; Dstyk; Dual serine/threonine and tyrosine protein kinase; dusty PK; Dusty protein kinase; DustyPK; HDCMD38P; KIAA0472; receptor interacting protein kinase 5; receptor-interacting serine/threonine-protein kinase 5; RIP5; RIP-homologous kinase; Ripk5; sgK496; Sugen kinase 496
Gene Symbols Dstyk
Host Species Rabbit
Immunogen Fusion protein of human DSTYK.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25778
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.