Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIPPLY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570769 Supplier Invitrogen™ Supplier No. PA570769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Horse: 79%; Human: 100%; Rat: 79%.

This gene encodes a protein similar to a zebrafish protein which acts as a transcriptional repressor in and is required for somite segmentation in zebrafish embryos. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPPLY1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RIPPLY1
Gene Accession No. Q0D2K3
Gene Alias protein ripply1; ripply transcriptional repressor 1; RIPPLY1; ripply1 homolog
Gene Symbols RIPPLY1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RIPPLY1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92129
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.