Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPPLY2 Monoclonal Antibody (OTI1B5), TrueMAB™, OriGene
SDP

Catalog No. 501672653
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2653 100 μL
50-254-2224 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-2653 Supplier OriGene Supplier No. TA501512
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RIPPLY2 encodes a nuclear protein that belongs to a novel family of proteins required for vertebrate somitogenesis. Members of this family have a tetrapeptide WRPW motif that is required for interaction with the transcriptional repressor Groucho and a carboxy-terminal Ripply homology domain/Bowline-DSCR-Ledgerline conserved region required for transcriptional repression. Null mutant mice die soon after birth and display defects in axial skeleton segmentation due to defective somitogenesis. Alternative splicing results in multiple transcript variants.

Specifications

Antigen RIPPLY2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RIPPLY2
Gene Accession No. Q5TAB7
Gene Alias C6orf159, SCDO6, dJ237I15.1
Gene Symbols RIPPLY2
Host Species Mouse
Immunogen Full length human recombit protein of human RIPPLY2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134701
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.