Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIPPLY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561521 Supplier Invitrogen™ Supplier No. PA561521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MENAGGAEGT ESGAAACAAT DGPTRRAGAD SGYAGFWRPW VDAGGKKEEE TPNHAAEAMP DGPGMTAASG KLYQFRHPV Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 43%.

RIPPLY2 encodes a nuclear protein that belongs to a novel family of proteins required for vertebrate somitogenesis. Members of this family have a tetrapeptide WRPW motif that is required for interaction with the transcriptional repressor Groucho and a carboxy-terminal Ripply homology domain/Bowline-DSCR-Ledgerline conserved region required for transcriptional repression. Null mutant mice die soon after birth and display defects in axial skeleton segmentation due to defective somitogenesis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPPLY2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RIPPLY2
Gene Accession No. Q5TAB7
Gene Alias C6orf159; dJ237I15.1; Protein ripply2; ripply transcriptional repressor 2; RIPPLY2; ripply2 homolog; SCDO6
Gene Symbols RIPPLY2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RIPPLY2. Recombinant protein control fragment (Product #RP-100308).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134701
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.