Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIPX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588846
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588846 Supplier Invitrogen™ Supplier No. PA588846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSALTPPTDM PTPTTDKITQ AAMETIYLCK FRVSMDGEWL CLRELDDISL TPDPEPTHED PNYLM; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell projection, Cytoplasm, Endomembrane system, Perikaryon, filopodium, growth cone, invadopodium, lamellipodium.

Implicated in the formation of a single axon by developing neurons. May inhibit the formation of additional axons by inhibition of PI3K in minor neuronal processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPX
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RUFY3
Gene Accession No. Q5FVJ0, Q9D394
Gene Alias 2810428M05Rik; 6330416M07Rik; AW455998; AW538594; D5Bwg0860e; KIAA0871; Protein RUFY3; rap2 interacting protein x; rap2-interacting protein x; RIPx; Rpipx; RUFY3; RUN and FYVE domain containing 3; RUN and FYVE domain-containing protein 3; Singar; SINGAR1; singar2; single axon-regulated protein; Single axon-regulated protein 1; single axon-related 1; ZFYVE30
Gene Symbols RUFY3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-65 of human RUFY3 (NP_0557761).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360921, 52822
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.