Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RISC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583448 Supplier Invitrogen™ Supplier No. PA583448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPVISIVDEL LEAGINVTVY NGQLDLIVDT MGQEAWVRKL KWPELPKFSQ LKWKALYSDP KSLETSAFVK SYKNLAFYWI LKAGHMVPSD QGDMALKMMR LVTQQ Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 85%.

The RNA-induced silencing complex (RISC) is a key player in the regulation of gene expression through RNA interference (RNAi) mechanisms. It operates by utilizing small RNA molecules-such as microRNAs (miRNAs), small interfering RNAs (siRNAs), and PIWI-interacting RNAs (piRNAs)-which are processed from double-stranded RNA precursors by enzymes like Drosha and Dicer in animals, and DICER-LIKE in plants. RISC is a versatile molecular complex consisting of Argonaute proteins as its core component, which guides the complex to the target RNA by complementary base pairing. Once assembled, RISC can recruit other proteins that mediate gene silencing by degrading or inhibiting the translation of the target RNA. This post-transcriptional regulation system is integral to numerous biological processes, allowing cells to control gene expression finely and respond to endogenous and exogenous signals. The functionality of RISC in gene silencing highlights its significance in the broader context of genetic regulation and has implications in therapies that target gene expression through RNAi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RISC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCPEP1
Gene Accession No. Q920A5, Q920A6, Q9HB40
Gene Alias 2410018F01Rik; 4833411K15Rik; AI957256; HSCP1; MSTP034; retinoid inducible serine carboxypeptidase; retinoid-inducible serine carboxypeptidase; retinoid-inducible serine caroboxypeptidase; retinoid-inducible serine caroboxypetidase; Risc; SCP1; SCPEP1; Serine carboxypeptidase 1; serine carboxypeptidase 1 precursor protein; serine caroboxypeptidase 1; UNQ265/PRO302
Gene Symbols SCPEP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCPEP1. Recombinant protein control fragment (Product #RP-93236).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114861, 59342, 74617
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.