Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Risdiplam
GREENER_CHOICESDP

Catalog No. 791650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-165-0 50 mg
79-161-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 79-165-0 Supplier Tocris Bioscience™ Supplier No. 7916/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective SMN2 gene splicing modifier

Risdiplam is a selective survival of motor neuron-2 (SMN2) pre-mRNA splicing modifier. It potently enhances SMN2 splicing in vitro (EC1.5X = 4 nM) and increases SMN protein in both brain (up to 100%) and muscle tissues (up to 49%) of transgenic mouse models of spinal muscular atrophy (SMA). Risdiplam dose-dependently increases the number of vesicular glutamate transporter 1 (vGlut1) proprioceptive inputs on motor neurons, the number of L3 to L5 motor neurons, the percentage of fully innervated neuromuscular junctions, and muscle size in the extensor digitorum longus muscles in SMN Δ7 mice.

Specifications

Chemical Name or Material 7-(4,7-Diazaspiro[2.5 ]oct-7-yl)-2-(2,8-dimethylimidazo[1,2-b]pyridazin-6-yl)-4 H-pyrido[1,2-a]pyrimidin-4-one
CAS 1825352-65-5
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C22H23N7O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.