Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Risedronic acid sodium salt, Thermo Scientific Chemicals

Catalog No. AC462930010
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115436-72-1
Molecular Formula C7H10NNaO7P2
Molecular Weight (g/mol) 305.09
MDL Number MFCD00867080,MFCD01706268,MFCD00867080
InChI Key DRFDPXKCEWYIAW-UHFFFAOYNA-M
IUPAC Name sodium hydrogen [1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethyl]phosphonate
SMILES [Na+].OC(CC1=CN=CC=C1)(P(O)(O)=O)P(O)([O-])=O

Specifications

Quantity 1 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: insoluble in ethanol
Formula Weight 305.1
Chemical Name or Material Risedronic acid sodium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.