Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530919 Supplier Invitrogen™ Supplier No. PA530919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Predicted reactivity: Mouse (97%), Zebrafish (87%), Xenopus laevis (87%), Pig (97%), Chicken (96%), Sheep (99%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Rit and its neuron-specific homologue, Rin, define a recently discovered subfamily of Ras-related GTPases. Rit and Rin are membrane-associated in spite of the fact that they lack a CAAX box or similar C-terminal lipidation motif. Rit and Rin display 64% amino acid sequence identity and share a unique nine amino acid effector domain (DPTIEDAYK) that is 100% conserved between the murine and human proteins. Although the effector domain sequences of Rit and Rin are very similar to that of Ras, Rit and Rin have been shown to interact with the known Ras-binding proteins RalGDS, Rlf and AF-6, but not the Raf kinases, RIN1 or the p110 subunit of PI3 kinase. For this reason, it has been suggested that Rit and Rin may play important roles in the regulation of signaling pathways distinct from those controlled by Ras.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RIT1
Gene Accession No. Q92963
Gene Alias GTP-binding protein Rit1; GTP-binding protein Roc1; MGC125864; MGC125865; NS8; Ras like without CAAX 1; ras-like protein expressed in many tissues; Ras-like without CAAX 1; ras-like without CAAX protein 1; RGD1559874; RIBB; Ric-like, expressed in many tissues; RIT; RIT1; ROC1; si:ch211-93a2.3
Gene Symbols RIT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RIT1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6016
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.