Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rit2 Antibody (OTI3F4), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273917FR
Description
Rit2 Monoclonal specifically detects Rit2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Rit2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
RIT2 | |
Full length recombinant protein of human Rit2 (NP_002921) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI3F4 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
GTP-binding protein Rit2, GTP-binding protein Roc2, Ras-like protein expressed in neurons, Ras-like without CAAX 2, Ras-like without CAAX protein 2, RIBA, Ric (Drosophila)-like, expressed in neurons, Ric-like, expressed in neurons, RIN, ROC2 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
6014.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction