Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIT2 Monoclonal Antibody (OTI3F4), TrueMAB™, OriGene
SDP

Catalog No. 501672875
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2875 100 μL
50-254-2041 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-2875 Supplier OriGene Supplier No. TA501757
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RIT2 belongs to the RAS (HRAS) superfamily of small GTPases.

Specifications

Antigen RIT2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3F4
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RIT2
Gene Accession No. Q5BJQ5, Q99578
Gene Alias RIBA, RIN, ROC2
Gene Symbols Rit2
Host Species Mouse
Immunogen Full length human recombit protein of human RIT2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291713, 490475, 6014
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.