Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIT2 Monoclonal Antibody (OTI4B5), TrueMAB™, OriGene
SDP

Catalog No. 502542084
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2084 30 μL
50-167-2823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2084 Supplier OriGene Supplier No. TA501704S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RIT2 Monoclonal antibody specifically detects RIT2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen RIT2
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4B5
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RIT2
Gene Accession No. Q99578
Gene Alias GTP-binding protein Rit2; GTP-binding protein Roc2; Ras like without CAAX 2; RAS-like protein expressed in neuron; ras-like protein expressed in neurons; Ras-like without CAAX 1; Ras-like without CAAX 2; ras-like without CAAX protein 2; RIBA; Ric-like, expressed in neurons; RIN; Rit1; RIT2; ROC2
Host Species Mouse
Immunogen Full length human recombinant protein of human RIT2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6014
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.