Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RITA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558654 Supplier Invitrogen™ Supplier No. PA558654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGPSKTEPGP AADSQKLSMG GLHSSRPLKR GLSHSLTHLN VPSTGHPATS APHTNGPQDL RPSTSGVTFR SPLVTSRARS VSISVPST Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 56%.

Tubulin-binding protein that acts as a negative regulator of Notch signaling pathway. Shuttles between the cytoplasm and the nucleus and mediates the nuclear export of RBPJ/RBPSUH, thereby preventing the interaction between RBPJ/RBPSUH and NICD product of Notch proteins (Notch intracellular domain), leading to down-regulate Notch-mediated transcription. May play a role in neurogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RITA1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RITA1
Gene Accession No. Q96K30
Gene Alias C12orf52; PSEC0043; RBPJ interacting and tubulin associated 1; RBPJ-interacting and tubulin-associated protein; RBPJ-interacting and tubulin-associated protein 1; RITA; RITA1
Gene Symbols RITA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RITA1. Recombinant protein control fragment (Product #RP-97761).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84934
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.