Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rituximab Recombinant Human Monoclonal Antibody (10F381 (rituximab))
GREENER_CHOICE

Catalog No. PIMA547773
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547773 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547773 Supplier Invitrogen™ Supplier No. MA547773
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

Specificity: CD20 (Human B-lymphocyte-restricted differentiation antigen, Bp35), is a hydrophobic transmembrane protein with a molecular weight of approximately 35 kDa located on pre-B and mature B lymphocytes, and most B-cell non-Hodgkin's lymphomas, but is not found on stem cells, pro-B cells, normal plasma cells or other normal tissues.

Rituximab (Rituxan) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with certain autoimmune diseases and types of cancer, such as non-Hodgkin's lymphoma, chronic lymphocytic leukemia, rheumatoid arthritis, idiopathic thrombocytopenic purpura, and pemphigus vulgaris. Rituximab is a chimeric monoclonal antibody against the protein CD20, which is primarily found on the surface of immune system B cells. When it binds to this protein it triggers cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rituximab
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 10F381 (rituximab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Fcgr3
Gene Accession No. P08101, P08508
Gene Alias IDEC-C2B8; Rituxan
Host Species Human
Immunogen Human lymphoblastoid cell line SB.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14130, 14131
Target Species Human, Rhesus Macaque, Cynomolgus Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.